Imprimer

Anti-FTO Rabbit Polyclonal Antibody (DyLight® 550)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-60935R
NOVUNB110-60935REA 709 EUR
NOVUNB110-60935R
Anti-FTO Rabbit Polyclonal Antibody (DyLight® 550)
Anticorps
Type: Primary
Antigen: FTO (fat mass and obesity associated)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR