Imprimer

Anti-Hepatic Sinusoidal Endothelial Cells Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: SE-1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-68095F
NOVUNB110-68095FEA 723 EUR
NOVUNB110-68095F
Anti-Hepatic Sinusoidal Endothelial Cells Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: SE-1]
Anticorps
Type: Primary
Antigen: Hepatic Sinusoidal Endothelial Cells
Clonality: monoclonal
Clone: SE-1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR