Imprimer

Anti-TNC Mouse Monoclonal Antibody (DyLight® 550) [clone: 4C8MS]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-68136REA 694 EUR
NOVUNB110-68136R
Anti-TNC Mouse Monoclonal Antibody (DyLight® 550) [clone: 4C8MS]
Anticorps
Type: Primary
Antigen: TNC (tenascin C)
Clonality: monoclonal
Clone: 4C8MS
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR