Imprimer

Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 488) [clone: FDO66Q]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-78644G
NOVUNB110-78644GEA 709 EUR
NOVUNB110-78644G
Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 488) [clone: FDO66Q]
Anticorps
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: monoclonal
Clone: FDO66Q
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR