Imprimer

Anti-SLC9A3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 14D5]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-81529H
NOVUNB110-81529HEA 711 EUR
NOVUNB110-81529H
Anti-SLC9A3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 14D5]
Anticorps
Type: Primary
Antigen: SLC9A3 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3)
Clonality: monoclonal
Clone: 14D5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR