Imprimer

Anti-SPP1 Mouse Monoclonal Antibody (DyLight® 680) [clone: 1B20]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-89062FR
NOVUNB110-89062FREA 667 EUR
NOVUNB110-89062FR
Anti-SPP1 Mouse Monoclonal Antibody (DyLight® 680) [clone: 1B20]
Anticorps
Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: monoclonal
Clone: 1B20
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR