Imprimer

Anti-ACSL1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-99585F
NOVUNB110-99585FEA 709 EUR
NOVUNB110-99585F
Anti-ACSL1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: ACSL1 (acyl-CoA synthetase long-chain family member 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR