Imprimer

Anti-TP53 Mouse Monoclonal Antibody (DyLight® 755) [clone: PAb 240]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB200-103IR
NOVUNB200-103IREA 667 EUR
NOVUNB200-103IR
Anti-TP53 Mouse Monoclonal Antibody (DyLight® 755) [clone: PAb 240]
Anticorps
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: PAb 240
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR