Imprimer

Anti-MYC Mouse Monoclonal Antibody (DyLight® 488) [clone: 9E11]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB200-108G
NOVUNB200-108GEA 723 EUR
NOVUNB200-108G
Anti-MYC Mouse Monoclonal Antibody (DyLight® 488) [clone: 9E11]
Anticorps
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 900000000000
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR