Imprimer

Anti-CDKN2A Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB200-111FEA 681 EUR
NOVUNB200-111F
Anti-CDKN2A Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR