Imprimer

Anti-NQO1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: A180]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB200-209APC
NOVUNB200-209APCEA 667 EUR
NOVUNB200-209APC
Anti-NQO1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: A180]
Anticorps
Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: monoclonal
Clone: A180
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR