Imprimer

Anti-RNA polymerase II Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4H8]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB200-598APCEA 696 EUR
NOVUNB200-598APC
Anti-RNA polymerase II Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4H8]
Anticorps
Type: Primary
Antigen: RNA polymerase II
Clonality: monoclonal
Clone: 4H8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR