Imprimer

Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB21-1101B
NOVUNB21-1101BEA 821 EUR
NOVUNB21-1101B
Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR