Imprimer

Anti-Neurokinin 1 Receptor Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB300-119PE
NOVUNB300-119PEEA 785 EUR
NOVUNB300-119PE
Anti-Neurokinin 1 Receptor Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorps
Type: Primary
Antigen: Neurokinin 1 Receptor
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR