Imprimer

Anti-NF1 Mouse Monoclonal Antibody (DyLight® 488) [clone: McNFn27a]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB300-153G
NOVUNB300-153GEA 637 EUR
NOVUNB300-153G
Anti-NF1 Mouse Monoclonal Antibody (DyLight® 488) [clone: McNFn27a]
Anticorps
Type: Primary
Antigen: NF1 (neurofibromin 1)
Clonality: monoclonal
Clone: McNFn27a
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Guinea pig, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR