Imprimer

Anti-CTDSPL Rabbit Polyclonal Antibody (DyLight® 650)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB300-231C
NOVUNB300-231CEA 681 EUR
NOVUNB300-231C
Anti-CTDSPL Rabbit Polyclonal Antibody (DyLight® 650)
Anticorps
Type: Primary
Antigen: CTDSPL (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR