Imprimer

Anti-SCARB1 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB400-101B
NOVUNB400-101BEA 737 EUR
NOVUNB400-101B
Anti-SCARB1 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR