Imprimer

Anti-ABCG8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB400-110H
NOVUNB400-110HEA 630 EUR
NOVUNB400-110H
Anti-ABCG8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Type: Primary
Antigen: ABCG8 (ATP-binding cassette, sub-family G (WHITE), member 8)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR