Imprimer

Anti-ABCF2 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB400-115 NB400-115SS
NOVUNB400-115EA 542 EUR
NOVUNB400-115 NOVUNB400-115SS
Anti-ABCF2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ABCF2 (ATP-binding cassette, sub-family F (GCN20), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR