Imprimer

Anti-Niemann-Pick type C1 Like-1 Rabbit Polyclonal Antibody (DyLight® 405)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB400-127V
NOVUNB400-127VEA 696 EUR
NOVUNB400-127V
Anti-Niemann-Pick type C1 Like-1 Rabbit Polyclonal Antibody (DyLight® 405)
Anticorps
Type: Primary
Antigen: Niemann-Pick type C1 Like-1
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR