Imprimer

Anti-THOP1 Mouse Monoclonal Antibody (DyLight® 755) [clone: 4D6]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB400-146IREA 642 EUR
NOVUNB400-146IR
Anti-THOP1 Mouse Monoclonal Antibody (DyLight® 755) [clone: 4D6]
Anticorps
Type: Primary
Antigen: THOP1 (thimet oligopeptidase 1)
Clonality: monoclonal
Clone: 4D6
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR