Imprimer

Anti-Niemann-Pick C1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB400-148F
NOVUNB400-148FEA 634 EUR
NOVUNB400-148F
Anti-Niemann-Pick C1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: Niemann-Pick C1
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR