Imprimer

Anti-MRP1 Mouse Monoclonal Antibody [clone: IU2H10]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB400-156EA 604 EUR
NOVUNB400-156 NOVUNB400-156SS
Anti-MRP1 Mouse Monoclonal Antibody [clone: IU2H10]
Anticorps
Type: Primary
Antigen: MRP1
Clonality: monoclonal
Clone: IU2H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR