Imprimer

Anti-MRP1 Mouse Monoclonal Antibody (DyLight® 488) [clone: IU2H10]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB400-156G
NOVUNB400-156GEA 667 EUR
NOVUNB400-156G
Anti-MRP1 Mouse Monoclonal Antibody (DyLight® 488) [clone: IU2H10]
Anticorps
Type: Primary
Antigen: MRP1
Clonality: monoclonal
Clone: IU2H10
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR