Imprimer

Anti-PCNA Mouse Monoclonal Antibody (DyLight® 350) [clone: PC10]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB500-106UV
NOVUNB500-106UVEA 723 EUR
NOVUNB500-106UV
Anti-PCNA Mouse Monoclonal Antibody (DyLight® 350) [clone: PC10]
Anticorps
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone: PC10
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR