Imprimer

Anti-TPX2 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB500-179 NB500-179SS
NOVUNB500-179EA 615 EUR
NOVUNB500-179 NOVUNB500-179SS
Anti-TPX2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TPX2 (TPX2, microtubule-associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR