Imprimer

Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 755) [clone: 60.11]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB500-238IR
NOVUNB500-238IREA 696 EUR
NOVUNB500-238IR
Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 755) [clone: 60.11]
Anticorps
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 60.11
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR