Imprimer

Anti-BBC3 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB500-261 NB500-261SS
NOVUNB500-261EA 542 EUR
NOVUNB500-261 NOVUNB500-261SS
Anti-BBC3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: BBC3 (BCL2 binding component 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR