Imprimer

Anti-PDPN Golden syrian hamster Monoclonal Antibody (DyLight® 488) [clone: 8.1.1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB600-1015G
NOVUNB600-1015GEA 680 EUR
NOVUNB600-1015G
Anti-PDPN Golden syrian hamster Monoclonal Antibody (DyLight® 488) [clone: 8.1.1]
Anticorps
Type: Primary
Antigen: PDPN (podoplanin)
Clonality: monoclonal
Clone: 8.1.1
Conjugation: DyLight® 488
Epitope:
Host: Golden Syrian Hamster
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR