Imprimer

Anti-CD34 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MEC 14.7]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB600-1071F
NOVUNB600-1071FEA 640 EUR
NOVUNB600-1071F
Anti-CD34 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MEC 14.7]
Anticorps
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: MEC 14.7
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR