Imprimer

Anti-DDIT3 Mouse Monoclonal Antibody (DyLight® 350) [clone: 9C8]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB600-1335UV
NOVUNB600-1335UVEA 667 EUR
NOVUNB600-1335UV
Anti-DDIT3 Mouse Monoclonal Antibody (DyLight® 350) [clone: 9C8]
Anticorps
Type: Primary
Antigen: DDIT3 (DNA-damage-inducible transcript 3)
Clonality: monoclonal
Clone: 9C8
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR