Imprimer

Anti-LC3B Rabbit Polyclonal Antibody (DyLight® 405)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB600-1384V
NOVUNB600-1384VEA 737 EUR
NOVUNB600-1384V
Anti-LC3B Rabbit Polyclonal Antibody (DyLight® 405)
Anticorps
Type: Primary
Antigen: LC3B
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR