Imprimer

Anti-IFITM3 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB600-1534
NOVUNB600-1534EA 431 EUR
NOVUNB600-1534
Anti-IFITM3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: IFITM3 (interferon induced transmembrane protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR