Imprimer

Anti-PRKCA Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: MC5]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB600-201PCP
NOVUNB600-201PCPEA 696 EUR
NOVUNB600-201PCP
Anti-PRKCA Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: MC5]
Anticorps
Type: Primary
Antigen: PRKCA (protein kinase C, alpha)
Clonality: monoclonal
Clone: MC5
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR