Imprimer

Anti-MDH1 / MOR2 Goat Polyclonal Antibody

Fournisseur: Everest Biotechnology
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

EVBIEB11994EA 310 EUR
EVBIEB11994
Anti-MDH1 / MOR2 Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: MDH1 / MOR2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR