Imprimer
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Fournisseur: Rockland Immunochemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ROCK610-4104EA 268 EUR
ROCK610-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Anticorps
Secondary Rabbit Anti-IgG F(ab')2 Reacts with Mouse
Recommended Dilutions: ELISA: 1:75000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Mouse