Imprimer

Anti-ACOX1 Rabbit Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 153CT43.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030738-HRPEA 720 EUR
USBI030738-HRP
Anti-ACOX1 Rabbit Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 153CT43.1.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ACOX1 (acyl-CoA oxidase 1, palmitoyl)
Clonality: Monoclonal
Clone: 153CT43.1.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR