Imprimer

Anti-ACTA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 337CT30.10.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030740-APCEA 720 EUR
USBI030740-APC
Anti-ACTA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 337CT30.10.1]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry 1:10-50

Type: Primary
Antigen: ACTA1 (actin, alpha 1, skeletal muscle)
Clonality: Monoclonal
Clone: 337CT30.10.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR