Imprimer

Anti-ACTB Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 137CT26.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030742-APEA 728 EUR
USBI030742-AP
Anti-ACTB Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 137CT26.1.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-4000; Immunochemistry1:50-100; Immunofluorescence: 1:100-500

Type: Primary
Antigen: ACTB (actin, beta)
Clonality: Monoclonal
Clone: 137CT26.1.1
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1 Kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR