Imprimer

Anti-ADA Mouse Monoclonal Antibody [clone: 608CT2.1.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030745EA 527 EUR
USBI030745
Anti-ADA Mouse Monoclonal Antibody [clone: 608CT2.1.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-8000

Type: Primary
Antigen: ADA (adenosine deaminase)
Clonality: Monoclonal
Clone: 608CT2.1.3
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR