Imprimer

Anti-AGT Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 656CT5.8.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030750-APEA 711 EUR
USBI030750-AP
Anti-AGT Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 656CT5.8.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: AGT (angiotensinogen (serpin peptidase inhibitor, clade A, member 8))
Clonality: Monoclonal
Clone: 656CT5.8.4
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR