Imprimer

Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 449CT13.4.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030754EA 495 EUR
USBI030754
Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 449CT13.4.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: AKT1S1 (AKT1 substrate 1 (proline-rich))
Clonality: Monoclonal
Clone: 449CT13.4.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR