Imprimer

Anti-AKT2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 148CT5.4.1.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030756-HRPEA 720 EUR
USBI030756-HRP
Anti-AKT2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 148CT5.4.1.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: Monoclonal
Clone: 148CT5.4.1.4
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR