Imprimer

Anti-ALDH3A2 Mouse Monoclonal Antibody (Biotin) [clone: 151CT1.3.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030765-BIOTINEA 711 EUR
USBI030765-BIOTIN
Anti-ALDH3A2 Mouse Monoclonal Antibody (Biotin) [clone: 151CT1.3.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:10-50; Immunochemistry: 1:50-100

Type: Primary
Antigen: ALDH3A2 (aldehyde dehydrogenase 3 family, member A2)
Clonality: Monoclonal
Clone: 151CT1.3.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR