Imprimer

Anti-AMBP Mouse Monoclonal Antibody [clone: 472CT11.5.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030770EA 527 EUR
USBI030770
Anti-AMBP Mouse Monoclonal Antibody [clone: 472CT11.5.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-800

Type: Primary
Antigen: AMBP (alpha-1-microglobulin/bikunin precursor)
Clonality: Monoclonal
Clone: 472CT11.5.4
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR