Imprimer

Anti-ANPEP Mouse Monoclonal Antibody (Biotin) [clone: 505CT12.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030775-BIOTINEA 711 EUR
USBI030775-BIOTIN
Anti-ANPEP Mouse Monoclonal Antibody (Biotin) [clone: 505CT12.1.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone: 505CT12.1.2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR