Imprimer

Anti-APCS Mouse Monoclonal Antibody [clone: 342CT9.4.7]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030776EA 527 EUR
USBI030776
Anti-APCS Mouse Monoclonal Antibody [clone: 342CT9.4.7]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-4000

Type: Primary
Antigen: APCS (amyloid P component, serum)
Clonality: Monoclonal
Clone: 342CT9.4.7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR