Imprimer

Anti-ATG5 Mouse Monoclonal Antibody (Biotin) [clone: 22CT1.11.4.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030779-BIOTINEA 711 EUR
USBI030779-BIOTIN
Anti-ATG5 Mouse Monoclonal Antibody (Biotin) [clone: 22CT1.11.4.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: ATG5 (autophagy related 5)
Clonality: Monoclonal
Clone: 22CT1.11.4.4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR