Imprimer

Anti-APOA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 464CT10.4.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030784-APCEA 711 EUR
USBI030784-APC
Anti-APOA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 464CT10.4.4]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: Monoclonal
Clone: 464CT10.4.4
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR