Imprimer

Anti-ARSA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 498CT24.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI030787-FITCEA 711 EUR
USBI030787-FITC
Anti-ARSA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 498CT24.1.2]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: ARSA (arylsulfatase A)
Clonality: Monoclonal
Clone: 498CT24.1.2
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR